Nunc™ Universal and Transport Containers with Caps
Nunc™ Universal and Transport Containers with Caps
Actual product may vary
Nunc™ Universal and Transport Containers with Caps
Nunc™ Universal and Transport Containers with Caps
Nunc™ Universal and Transport Containers with Caps
Fisherbrand
Nunc™ Universal and Transport Containers with Caps
Store and transport blood, urine, fecal and other specimens safely using these universal and transport containers with caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialSterilityLabelIncludes
FKS10779081PolystyreneSterileLabeledSpoon
FKS10356041Polystyrene (High Impact)Non-sterileNo LabelNo Spoon
Catalog number FKS10779081
Price (SGD)
-
Material:
Polystyrene
Sterility:
Sterile
Label:
Labeled
Includes:
Spoon
Safely store and transport blood, urine, fecal and other specimens using Thermo Scientific™ Nunc™ Universal and Transport Containers with Caps. These containers are designed with inner and outer containers of high mechanical strength. Caps are designed to avoid droplets adhering to the surface, reducing risk of aerosols. Nunc Labware Products are made from high purity resins, and molded using our state-of-the-art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.
Universal Containers
  • Working volume: 25mL
  • Outer case available in transparent or opaque material
  • Made of polystyrene with polyethylene caps (except 374799 which has polystyrene body with polyethylene, high-impact polystyrene and low density polystyrene)
  • Available sterile or nonsterile, and with or without a spoon for handling of solid and semi-solid samples
  • Some available with writing area
  • Max. RCF tested: 4000
Transport Container
  • Polypropylene with polyethylene cap or high impact polystyrene with polyethylene
Specifications
Product TypeUniversal Container with Spoon
Closure IncludedYes
MaterialPolystyrene
SterilitySterile
Capacity (Metric)25 mL
Certifications/ComplianceTransport and Universal Containers have passed the 95 kPa test described in UN packaging instructions 602/650 for transport of infectious substances and diagnostic specimens. Universal containers are CE marked.
Closure MaterialPolyethylene
ColorClear
DescriptionW/spoon; Sterile; PS; Writing area
IncludesSpoon
LabelLabeled
No. per Case300/Cs.
No. per Pack50/Pk.
RCF Rating4000 x g
Unit SizePK