HDPE, Jerry Can
HDPE, Jerry Can
Actual product may vary
Fisherbrand
HDPE, Jerry Can
Canister for storage or transport. Canister with UN transport approval and tamper-evident seal.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Dimensions (L x W x H)
FKS107705935 L190 x 145 x 251 mm
FKS1061369320 L290 x 246 x 385 mm
Catalog number FKS10770593
Price (SGD)
36.00
EA
Add to cart
Capacity (Metric):
5 L
Dimensions (L x W x H):
190 x 145 x 251 mm
Price (SGD)
36.00
EA
Add to cart
  • For storage or transport
  • HDPE
  • With UN approval for transport
  • Screw lid without drain valve is included, screw lid with drain valve available as accessory
Specifications
AutoclavableNot Autoclavable
Capacity (English)1.3 gal.
Capacity (Metric)5 L
Certifications/ComplianceUN approval 3H1/X1.9/250
Closure Size1427-9105, Orig. Screw Cap DIN50
Closure TypeDIN 50
ColorTransparent
Diameter (Metric) Inner Neck40.5 mm
Dimensions (L x W x H)190 x 145 x 251 mm
Dimensions (L x W)190 x 145 mm
For Use With (Application)For Storage or Transport
Height (Metric) With Closure251 mm
IncludesScrew lid
Length (Metric)190 mm
MaterialHDPE
Mouth StyleS50 Thread
No. per Pack1
Product TypeJerry Can
ShapeSquare
SterilityNon-sterile
Width (Metric)145 mm
Unit SizeEA