Stainless-Steel Test Sieves, 200 Dia. x 50 mm H, Pore sizes 10 mm and Smaller, ISO 3310/1
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Stainless-Steel Test Sieves, 200 Dia. x 50 mm H, Pore sizes 10 mm and Smaller, ISO 3310/1
Use for separation, fractioning, or particle size determination. These stainless-steel test sieves have an internal height of 50 mm and diameter of 200 mm. Available in a wide range of pore sizes between 1mm and 10 mm.
Have Questions?
Change view
Catalog Number
Pore Size
FKS10770273
5.6 mm
FKS10741433
3.35 mm
FKS10116601
4 mm
FKS10288781
2 mm
FKS10536222
1 mm
5 Options
Catalog number FKS10770273
Price (SGD)
509.00
PK
Add to cart
Pore Size:
5.6 mm
Price (SGD)
509.00
PK
Add to cart
Stainless-steel frame with wire gauze
Diameter: 200 mm
Height: 50 mm
ISO 3310/1
Specifications
Quantity1 Each
Pore Size5.6 mm
TypeTest Sieve
Certifications/ComplianceISO 3310/1. Main Series R’10 according to DIN 323
IncludesCertificate of compliance with ISO 3310/1 or NFX 11-504
Height (Metric)50mm
For Use With (Application)For separation, fractioning, particle size determination