Vivaspin™ 6 Centrifugal Concentrators
Vivaspin™ 6 Centrifugal Concentrators
Actual product may vary
Fisher Catalogue
Vivaspin™ 6 Centrifugal Concentrators
Achieve fast sample concentration and high recovery. Sartorius Vivaspin™ 6 Centrifugal Concentrators are the only device of its size that can process 6mL samples. They feature twin vertical membranes and built-in dead stop pockets. Available with PES membranes in a wide range of molecular weight cut-offs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMWCOQuantity
FKS107674615 kDa25/Pk.
FIS14-558-46310 kDa25/Pk.
Catalog number FKS10767461
View Price:Sign in to see your account pricing. Need an account? Register with us today.
MWCO:
5 kDa
Quantity:
25/Pk.
Developed to offer increased volume flexibility and performance
  • Compact tubes can process up to an impressive 6mL initial volume
  • Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
  • Universal rotor compatibility:
    • Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
Fast, safe and easy concentration
  • Features premium, high-throughput PES membranes
  • Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Nearly complete concentrate recovery of over 90%
  • Remaining volume is easy to see with the printed graduations and clear housing
  • Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate
Specifications
DisposableSingle-use
Filter TypeUltrafiltration
For Use With (Equipment)Centrifuge
Material (Membrane)PES
MWCO5 kDa
Sample Volume6 mL
SterilityNon-sterile
Filtration Area2.58 cm2
Housing MaterialPolycarbonate
Quantity25/Pk.
Volume (Metric)6 mL
Length (Metric)122 mm
Unit SizePK