Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) |
---|---|
FKS10761411 | 600 mL |
1201-0050 | 50 mL |
FIS02-591-10B | 100 mL |
1201-0150 | 150 mL |
FIS02-591-10C | 150 mL |
FKS10340481 | 150 mL |
1201-0250 | 250 mL |
FIS02-591-10D also known as FIS 02-591-10D | 250 mL |
FKS10547882 | 250 mL |
FIS02-591-10E | 400 mL |
1201-0600 | 600 mL |
FIS02-591-10F | 600 mL |
1201-1000 | 1000 mL |
FIS02-591-10G | 1000 mL |
1201-2000 | 2000 mL |
FIS02-591-10H | 2000 mL |
FIS02-591-10J | 4000 mL |
Carry out precise and efficient pouring with Thermo Scientific™ Nalgene™ Polypropylene Griffin Low-Form Beakers, featuring a single-spout design and easy-to-read silk-screened graduations. Rugged, reusable, construction that will last for years under typical lab conditions. Made only from high quality, laboratory-grade plastic materials for dependably low leachables and extractables. Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:
Ordering Information: Available in a size variety pack using Cat. No. 1201-1234
Recycling Information: Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene labware at end of product life in communities where polypropylene can be recycled.
Nalgene beakers and graduated cylinders are delivered in corrugate cartons made from 100% post-consumer recycled material. Packaging bags are made from LDPE and are readily recyclable once the label is removed. Please continue the cycle and recycle your cartons and bags.