Filter volumes up to 2mL with these 2mL polypropylene centrifugal filters
Have Questions?
Change view
Catalog Number
Material (Membrane)
Pore Size
FKS10753016
PVDF
0.2 μm
FKS10201345
Cellulose Acetate
0.22 μm
FKS10477254
Nylon
0.2 μm
FKS10077344
PTFE
0.2 μm
4 Options
Catalog number FKS10753016
Price (SGD)
527.00
EA
Add to cart
Material (Membrane):
PVDF
Pore Size:
0.2 μm
Price (SGD)
527.00
EA
Add to cart
Filter volumes up to 2mL with these Thermo Scientific™ National 2mL Nonsterile Centrifugal Filters. The filters feature a virgin polypropylene filter housing with tapered 5mL, capped receiver tube, and can be used with any benchtop or floor model laboratory centrifuge.
5,000xG maximum centrifugal force
Nonsterile
Bulk packed
Specifications
DescriptionPVDF Membrane; 0.2µm Pore Size
For Use With (Equipment)Benchtop or Floor Model Centrifuge