1x1 image pixel for data collection
Test Sieves - 14 Mesh or Greater
Test Sieves - 14 Mesh or Greater
Actual product may vary
Fisherbrand
Test Sieves - 14 Mesh or Greater
Stainless steel with wire gauze
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore Size
FKS10752003212 μm
FIS08-418-131 mm
Catalog number FKS10752003
Price (SGD)
612.00
EA
Add to cart
Pore Size:
212 μm
Price (SGD)
612.00
EA
Add to cart
Specifications
Depth (English)2 in.
Depth (Metric)5.08 cm
Pore Size212 μm
TypeTest Sieve
Certifications/ComplianceASTM E11
Diameter (English) Outer8 in.
MaterialStainless Steel
Tyler Standard70 Mesh
U.S. StandardNo. 70
For Use With (Application)Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc.
Diameter (Metric) Outer8 in.
Mesh MaterialStainless Steel
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link