Stomacher™ Lab Blender Bags
Stomacher™ Lab Blender Bags
Actual product may vary
Fisherbrand
Stomacher™ Lab Blender Bags
The Stomacher™ Bag range has been designed to fulfill all applications from sample blending, straining and storage, including bags designed for extensive blending and small samples for life science applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeCapacity (Metric)No. per Pack
FKS10746855Lab Blender Bags80 to 400 mL200
FIS14-285-25Lab Blender Bags80 to 400 mL500
FIS14-285-24Lab Blender Bags80 to 400 mL500
FKS10658165Lab Blender Bags80 to 400 mL200
FIS14-285-9Lab Blender Bags80 to 400 mL250
FIS14-285-20Lab Blender Bags80 to 400 mL250
FKS10547394Lab Blender Bags80 to 400 mL250
Catalog number FKS10746855
Price (SGD)
445.00
PK
Add to cart
Product Type:
Lab Blender Bags
Capacity (Metric):
80 to 400 mL
No. per Pack:
200
Price (SGD)
445.00
PK
Add to cart
  • Seward Stomacher bag range is made from high-quality food-grade virgin LDPE granule extruded to 65μm film thickness
  • Heavy-duty bags also available for large volume sampling 80μm film thickness; Sterile (gamma-irradiated) polyethylene bags; Closure bags feature wire closure strip across bag top for short term storage.
  • Strainer bags have inner plastic mesh to make it easy to pipette from samples that produce a large amount of debris
  • Single-chamber bags for use in Lab Blenders; internal sterility assured with proprietary tear-off sterile barrier top on sachets
  • Highly transparent and puncture-resistant virgin polyethylene extruded tube material
  • Simply tear the perforated end along the top of the sachet, remove the bag and open using Seward bag opener, insert the material to be analyzed and processed
  • Bags available for sample size 25ul - 4500ml
Specifications
DescriptionRound bottom; with Strainer
AutoclavableNot Autoclavable
Capacity (Metric)80 to 400 mL
Closure TypeNone
ColorClear
DisposableSingle-use
For Use With (Application)To prepare, store and transport samples in microbiology sample preparation
For Use With (Equipment)Stomacher 400 and all other 400ml paddle blenders
MaterialPolyethylene
SterilitySterile
Thickness (Metric)65 μm
No. per Pack200
Sterilization MethodGamma Irradiated
Dimensions (L x W)305mm x 177mm
Length (Metric)305 mm
Width (Metric)177 mm
Product TypeLab Blender Bags
Unit SizePK