Short-Term Detector Tubes: Benzene
Short-Term Detector Tubes: Benzene
Actual product may vary
Fisherbrand
Short-Term Detector Tubes: Benzene
Detector tubes
Have Questions?
Catalog NumberComponents MeasuredTube TypeMeasurement RangeHumidity Range
FKS10743263BenzeneBenzene 0.5/c0.5 to 101 to 30mg/L
Catalog number FKS10743263
Price (SGD)
692.00
PK
Add to cart
Components Measured:
Benzene
Tube Type:
Benzene 0.5/c
Measurement Range:
0.5 to 10
Humidity Range:
1 to 30mg/L
Price (SGD)
692.00
PK
Add to cart
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten. Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Specifications
Components MeasuredBenzene
Humidity Range1 to 30mg/L
Measurement Range0.5 to 10
No. of Strokes20
Quantity5
Response Time20 min.
Temperature Range (English)41°F to 104°F
Temperature Range (Metric)5° to 40°C
Tube TypeBenzene 0.5/c
Unit SizePK