Check Order Status
Set of 7 Adapters (Guko)
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Set of 7 Adapters (Guko)
Kartell™ Set of 7 Adapters (Guko) offers support to a Buchner Funnel and includes adapters with a variety of dimensions.
Have Questions?
Contact Us
Catalog Number
Type
FKS10737862
Adapters
Catalog number
FKS10737862
Price (SGD)
96.00
EA
Estimated availability date
01-Oct-2025
Add to cart
Type:
Adapters
Price (SGD)
96.00
EA
Add to cart
Supports for Buchner Funnels
Constructed of neoprene rubber.
Dimensions for adaptor 1:
Exterior dia. 21 x 11mm, Interior dia. 17 x 7mm, Height 21mm
Dimensions for adaptor 2:
Exterior dia. 27 x 16mm, Interior dia. 22 x 11mm, Height 22mm
Dimensions for adaptor 3:
Exterior dia. 37 x 22mm, Interior dia. 31 x 16mm, Height 25mm
Dimensions for adaptor 4:
Exterior dia. 46 x 29mm, Interior dia. 39 x 22mm, Height 29mm
Dimensions for adaptor 5:
Exterior dia. 58 x 38mm, Interior dia. 50 x 30mm, Height 35mm
Dimensions for adaptor 6:
Exterior dia. 69 x 45mm, Interior dia. 60 x 36mm, Height 40mm
Dimensions for adaptor 7:
Exterior dia. 86 x 57mm, Interior dia. 75 x 46mm, Height 45mm
Specifications
Color
Gray
For Use With (Equipment)
Buchner Funnel
Quantity
7 Adapters
Material
Neoprene
Type
Adapters
Unit Size
EA
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link