Grade 913 Lens Cleaning Tissue
Grade 913 Lens Cleaning Tissue
Actual product may vary
Fisherbrand
Grade 913 Lens Cleaning Tissue
Used for cleaning of optical lenses.
Have Questions?
Catalog number FKS10737643
Price (SGD)
160.00
PK
Add to cart
Price (SGD)
160.00
PK
Add to cart
  • Very thin paper
  • Made with 100% Manilla fibers
  • Not fluffy
  • Weight: 12g/m2
Application
  • Cleaning of various optical lens
  • Cleaning of optical fibers filaments
Specifications
Product TypeLens Cleaning Tissue
For Use With (Application)Cleaning of various optical lens (objective, microscopes binoculars, magnifying Glass, Glasses, etc.), cleaning of optical fibers filaments
MaterialManilla Fibers
Unit SizePK