1x1 image pixel for data collection
Big Squid Magnetic Stirrer
Big Squid Magnetic Stirrer
Actual product may vary
Fisherbrand
Big Squid Magnetic Stirrer
A magnetic stirrer with glass surfaces and different motives, providing outstanding chemical resistance.
Have Questions?
Catalog NumberModel
FKS10734007Big Squid White
Catalog number FKS10734007
Price (SGD)
-
Model:
Big Squid White
  • Digital speed display, LED
  • Electronically controlled motor for more capacity
  • Higher speed range from 0 to 2.500rpm
  • Maximum stirring quantity of 1.5L
  • Outstanding chemical resistance due to glass top and synthetic bottom made of TPC-ET
  • Recyclable materials
Specifications
Speed0 to 2500 rpm
Stirring Positions1
Top Plate MaterialGlass
Voltage100/240 V
Wattage4 w
Height (Metric)45 mm
Top Plate Surface Area (Metric)115 x 115 mm
Width (Metric)185 mm
Amperage300 mA
Frequency50/60 Hz
ModelBig Squid White
Motor TypeElectronically Controlled
Speed ControlTurning knob
Temperature Range (Metric)5°C to 40°C
TypeBig Squid White Magnetic Stirrer
Weight (Metric)0.7 kg
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection