Nalgene™ Polypropylene, Heavy-Duty Vacuum Carboy with Handle
Nalgene™ Polypropylene, Heavy-Duty Vacuum Carboy with Handle
Actual product may vary
Fisherbrand
Nalgene™ Polypropylene, Heavy-Duty Vacuum Carboy with Handle
For Use With: Ideal for the most extreme service conditions; Autoclavable; Closure Material: Polypropylene; Color: White
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1072868110 L
FIS02-960-1410 L
FIS02-960-1520 L
FKS1027430120 L
Catalog number FKS10728681
Price (SGD)
364.00
EA
Add to cart
Capacity (Metric):
10 L
Price (SGD)
364.00
EA
Add to cart
Ideal for the most extreme service conditions, the Thermo Scientific™ Nalgene™ Polypropylene Heavy Duty Vacuum Carboy is useful as a vacuum trap and will hold a full vacuum up to eight hours. Constructed of thick-walled natural polypropylene to provide added strength, it is supplied with a white 83B PP closure with thermoplastic elastomer (TPE) gasket. Customization options available for fluid transfer applications. Fit perfectly with a variety of optional 83B closures with standard tubing sets (such as Nalgene™ Top Works™, Nalgene™ Filling/Venting) and custom tubing set designs.
  • Autoclavable when closure threads are disengaged and vessel is empty
  • Not rated for pressure use
Ordering Information:
  • For closure options in vacuum applications, use with Nalgene Quick Filling/Venting Closures (Cat. No. 2158), or Nalgene Filling/Venting Closures (Cat. No. 2162)
  • Additional accessories include Nalgene Barbed Bulkhead Fittings (Cat. No. 6149)
  • For 1 to 5 L sizes, please refer to Nalgene Heavy Duty Vacuum Bottles (Cat. No. 2126)
Specifications
AutoclavableAutoclavable
Capacity (English)2.6 gal.
Capacity (Metric)10 L
Closure MaterialPolypropylene
Closure Size83B
ColorWhite
DescriptionHeavy-Duty Vacuum Carboy
Diameter (Metric) Outer250 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
IncludesHandles
MaterialPolypropylene
No. per Case6/Cs.
No. per Pack1 Ea.
Product TypeCarboy with Handle
SterilityNon-sterile
Volume (English) Brim3 gal.
Volume (Metric) Brim12 L
Unit SizeEA