1x1 image pixel for data collection
Azlon™ LDPE Specimen Container with Hinged Cap
Azlon™ LDPE Specimen Container with Hinged Cap
Actual product may vary
Fisherbrand
Azlon™ LDPE Specimen Container with Hinged Cap
Unbreakable and lightweight specimen containers. AzlonTM LDPE specimen containers with hinged caps are perfect for many laboratory applications.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FKS1071746510 mL28 mm32 mm
Catalog number FKS10717465
Price (SGD)
82.00
PK
Add to cart
Capacity (Metric):
10 mL
Diameter (Metric):
28 mm
Height (Metric):
32 mm
Price (SGD)
82.00
PK
Add to cart
  • Containers with captive hinged cap suitable for many applications
  • Lightweight and unbreakable in normal use
  • Can withstand temperatures up to 80°C
Specifications
Product TypeSpecimen Container
Closure IncludedYes
MaterialLDPE
Capacity (Metric)10 mL
Closure TypeHinged Cap
Diameter (Metric)28 mm
Height (Metric)32 mm
No. per Pack10/Pk.
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link