Nalgene™ Polypropylene, Wide-Mouth Carboy with Handle
Nalgene™ Polypropylene, Wide-Mouth Carboy with Handle
Actual product may vary
Fisherbrand
Nalgene™ Polypropylene, Wide-Mouth Carboy with Handle
For Use With: Store and transport solids, powders and liquids; Autoclavable; Closure Material: Polypropylene; Color: White
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1071269210 L
FIS02-961-65A10 L
FIS02-961-65C20 L
Catalog number FKS10712692
Price (SGD)
321.00
EA
Add to cart
Capacity (Metric):
10 L
Price (SGD)
321.00
EA
Add to cart
Store and transport solids, powders and liquids using Thermo Scientific™ Nalgene™ Wide-Mouth Autoclavable Polypropylene Carboys with Handles. These durable natural polypropylene (PP) carboys feature a convenient wide-mouth opening to promote ease of cleaning and dispensing. The wide-shoulder handles enable easy carrying and pouring, even with gloved hands.
  • Graduated to contain in 1 gal. and 5 L increments for ease of use
  • Wide-mouth opening can accommodate overhead mixers
  • Autoclavable
  • Leakproof† white polypropylene 100 mm screw closures
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
AutoclavableAutoclavable
Capacity (English)2.6 gal.
Capacity (Metric)10 L
Closure MaterialPolypropylene
Closure Size100-415 mm
ColorWhite
DescriptionAutoclavable Wide-Mouth Carboy with Handles
Diameter (Metric) Outer250 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Storage, Transportation ​
GraduationsGraduated to contain in 5 L/1 gal. increments
Height (Metric) With Closure343 mm
IncludesHandles
MaterialPolypropylene
Mouth StyleWide
No. per Case6/Cs.
No. per Pack1 Ea.
Product TypeCarboy with Handle
SterilityNon-sterile
Volume (English) Brim3 gal.
Volume (Metric) Brim12 L
Unit SizeEA