Amersham™ CyDye™ Fluorescent Nucleotides
Actual product may vary
Fisherbrand
Amersham™ CyDye™ Fluorescent Nucleotides
Label DNA and RNA probes. Cytiva Life Sciences™ Amersham™ CyDye™ Fluorescent Nucleotides are optimized for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.
Have Questions?
Catalog NumberProduct TypeQuantityColor
FKS10712257Cy3-dUTP25 nmolOrange
Catalog number FKS10712257
Price (SGD)
-
Product Type:
Cy3-dUTP
Quantity:
25 nmol
Color:
Orange
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.
  • Low nonspecific binding
  • High photostability
  • High water solubility
  • pH insensitive
Specifications
DescriptionCyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy dye labelled nucleotides exhibit the following properties:
pH7
Molecular Weight (g/mol)765.95 (Monofunction), 949.11 (Bisfunction)
Product TypeCy3-dUTP
ReagentsFluorescent Labeling Reagents
Wavelength570nm
Quantity25 nmol
ColorOrange
For Use With (Application)Genomic and Protein research, labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping and gene mapping
Unit SizePack
Contents & Storage
-15°C to -30°C