Polypropylene Tall Form Measuring Jugs
Polypropylene Tall Form Measuring Jugs
Actual product may vary
Fisherbrand
Polypropylene Tall Form Measuring Jugs
Manufactured from polypropylene for high thermal resistance. Kartell™ Measuring Jugs are transparent with permanent molded graduations for accurate measurements.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS107075412000 mL50/1mL
Catalog number FKS10707541
Price (SGD)
448.00
PK
Add to cart
Capacity (Metric):
2000 mL
Graduations:
50/1mL
Price (SGD)
448.00
PK
Add to cart
  • Convenient integral spout and handle allow easy dispensing
  • Autoclavable
  • Suitable for foodstuff
  • Tall form saves space in the laboratory
Specifications
Capacity (Metric)2000 mL
ColorTransparent
For Use With (Application)Foodstuff
FormTall form
MaterialPolypropylene
Graduations50/1mL
AutoclavableAutoclavable
GraduatedYes
Height (Metric)213 mm
Product TypeMeasuring Jug
Diameter (Metric)149 mm
Unit SizePK