Polypropylene Round Bottom Deep Well Plates
Polypropylene Round Bottom Deep Well Plates
Actual product may vary
Fisherbrand
Polypropylene Round Bottom Deep Well Plates
Accelerate your manual and automated work processes. Eppendorf™ Polypropylene Round Bottom Deep Well Plates combine a new well design with a sophisticated color coding system for ease of reading and orientation. Well geometry allows high speed pipetting without splashing, excellent mixing and high speed centrifugation.
Have Questions?
Catalog NumberColorTotal Volume (Metric) Well
FKS10700604Blue2000 μL
Catalog number FKS10700604
Price (SGD)
1,192.00
PK
Add to cart
Color:
Blue
Total Volume (Metric) Well:
2000 μL
Price (SGD)
1,192.00
PK
Add to cart
  • Recovermax - concaved round bottom well shape with smoothed edges
  • Optitrack - color framed plates with alphanumeric labelling
  • High chemical resistance
  • G-safe - highest centrifugation stability
  • LoBind - free of surface coating e.g., silicone
  • Free from DNA, DNase, RNase and PCR inhibitor (PCR clean)
  • LoBind Protein - minimum protein loss (less than 3% BSA 1μg/mL)
  • LoBind DNA - less than 1% loss of DNA/RNA at low concentrations and critical conditions (high salt, short DNA fragments)
  • Autoclavable
  • Optionally available with barcode
  • Fulfill all current SBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines
Specifications
BarcodeIncludes Barcode
Certifications/ComplianceSBS, IVD, EN ISO 9001/2/3 and EN 13485 guidelines
FormatStandard
Array8 x 12
AutoclavableAutoclavable
ColorBlue
MaterialPolypropylene
No. of Wells96
SterilityNon-sterile
Total Volume (Metric) Well2000 μL
Well ShapeRound Bottom (U-well)
Dnase FreeYes
For Use With (Application)Sample storage and preparation, bacteria and yeast cultivation, DNA and RNA isolation with high throughput
Quantity20/Pk.
Unit SizePK
Contents & Storage
-86°C