Azlon™ HDPE, Narrow-Neck Aspirator Bottle
Azlon™ HDPE, Narrow-Neck Aspirator Bottle
Actual product may vary
Fisherbrand
Azlon™ HDPE, Narrow-Neck Aspirator Bottle
For Use With: For bench top dispensing of common laboratory liquids/distilled water etc.; Color: White
Have Questions?
Catalog number FKS10696105
Price (SGD)
323.00
EA
Add to cart
Price (SGD)
323.00
EA
Add to cart
For Use With: For bench top dispensing of common laboratory liquids/distilled water etc.; Color: White
  • Raised graduations and translucent color
  • Smooth operating leak-proof stopcocks
  • Sturdy carry handle provides ease of transportation
  • Screw cap for secure storage
  • Conforms to USFDA guidelines for food contact
Specifications
Capacity (English)2.6 gal.
Capacity (Metric)10 L
ColorWhite
Diameter (Metric) Inner Neck49 mm
For Use With (Application)For bench top dispensing of common laboratory liquids/distilled water etc.
Height (Metric) With Closure380 mm
IncludesClosure
MaterialHDPE
Neck StyleNarrow
Product TypeAspirator Bottle
ShapeOctagonal
Unit SizeEA