1x1 image pixel for data collection
Autoclavable Safe-Lock Micropestle
Product Image
Fisherbrand
Autoclavable Safe-Lock Micropestle
Use this pestle for resuspending pellets. Eppendorf™ Autoclavable Safe-Lock Micropestles feature two sides for optimal fit in 1.5 and 2.0mL vessels.
Have Questions?
Catalog number FKS10683001
Price (SGD)
287.00
PK
Add to cart
Price (SGD)
287.00
PK
Add to cart
Use this pestle for resuspending pellets. Eppendorf™ Autoclavable Safe-Lock Micropestles feature two sides for optimal fit in 1.5 and 2.0mL vessels.
  • Autoclavable (121°C, 20 min.)
  • For use with 1.5 and 2.0mL vessels
  • Available in packs of ten
Specifications
AutoclavableAutoclavable
Product TypeMicropestle
For Use With (Application)For 1.5mL and 2mL Safe-Lock tubes and 1.7mL Safe-Twist screw cap tubes
Unit SizePK
Share catalog number, name or link