Haemocytometer Cover Glasses, Boro 3.3
Haemocytometer Cover Glasses, Boro 3.3
Actual product may vary
Haemocytometer Cover Glasses, Boro 3.3
Manufactured from borosilicate glass and complies with DIN ISO 8255 standards. BRAND™ Borosilicate Glass Haemacytometer Cover Glasses differ from ordinary cover glasses by their plain ground and polished surface.
Have Questions?
Catalog NumberLength (Metric)Width (Metric)
FKS1066397220 mm26 mm
Catalog number FKS10663972
Price (SGD)
896.00
PK
Add to cart
Length (Metric):
20 mm
Width (Metric):
26 mm
Price (SGD)
896.00
PK
Add to cart
  • Borosilicate glass, DIN ISO 8255.
  • Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Flatness tolerance ± 3 μm.
  • They differ from ordinary cover glasses by their plain ground and polished surface.
  • Packing: 2 cover glasses in a plastic bag, separated by tissue paper.
Specifications
Product TypeHemacytometer Coverslip
MaterialBorosilicate Glass
Length (Metric)20 mm
Width (Metric)26 mm
Thickness (Metric)0.4 mm
Dimensions (L x W)20 x 26 mm
Unit SizePK