DURAN™ Separating Funnel, conical shape, with PTFE key
DURAN™ Separating Funnel, conical shape, with PTFE key
Actual product may vary
DURAN™ Separating Funnel, conical shape, with PTFE key
DURAN™ Separating Funnel, conical shape, with PTFE key
Fisherbrand
DURAN™ Separating Funnel, conical shape, with PTFE key
The separating funnel enables the separation of immiscible liquids (liquid-liquid extraction).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS10658013250 mL
FKS10426332500 mL
FKS102435121000 mL
Catalog number FKS10658013
Price (SGD)
353.00
EA
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
353.00
EA
Add to cart
A separating funnel has the shape of a pointed flask with a drain cock at its tip. The separating funnel is used for the liquid-liquid extraction of two immiscible liquids (substance separation process) and can thus also be used for cleaning processes. The liquid with the higher density collects in the lower part of the vessel and can be drained via the drain cock. The printed scale allows an exact volume determination.
  • Conical shape
  • With PTFE key
  • Fired-on, highly durable white print
  • Easy-to-read scale with graduation marks to estimate the content
Specifications
Capacity (Metric)250 mL
Diameter (Metric) Stem10 mm
MaterialBoro 3.3, PTFE, Polypropylene
AutoclavableAutoclavable
For Use With (Application)Phase separation; Liquid-liquid extraction; Measuring liquid amounts
Quantity1 Ea.
Unit SizeEA