Centricon™ Plus-70 Centrifugal Filters
Centricon™ Plus-70 Centrifugal Filters
Actual product may vary
Fisherbrand
Centricon™ Plus-70 Centrifugal Filters
15 mL to 70 mL volume
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMWCO
FKS10654923100 kDa
FKS1029606210 kDa
FKS1004200230 kDa
Catalog number FKS10654923
Price (SGD)
1,389.00
PK
Add to cart
MWCO:
100 kDa
Price (SGD)
1,389.00
PK
Add to cart
The Centricon™ Plus-70 centrifugal filter is a disposable, single-use device designed for rapid processing of aqueous biological solutions in volumes ranging from 15 to 70 mL. It is compatible with swinging-bucket centrifuges only. The Centricon Plus-70 device can concentrate most 70 mL solutions down to 350 μL in just 25 minutes (some solutions may take slightly longer).•90% typical recovery •50X to 200X concentration •Low hold-up volume •Polypropylene housing minimizes binding •Deadstop prevents spinning to dryness •Ready to use. Nothing to assemble Applications:•Concentrating and desalting chromatography column eluates •Concentrating monoclonal antibodies •Concentrating proteins or viruses from culture supernatants •Clarifying tissue homogenates and cell lysates •Buffer exchange or diafiltration
Specifications
DisposableSingle-use
Material (Membrane)Ultracel-PL
MWCO100 kDa
SterilityNon-sterile
For Use With (Application)Concentrating and desalting chromatography column eluates; Concentrating monoclonal antibodies; Concentrating proteins or viruses from culture supernatants; Clarifying tissue homogenates and cell lysates; Buffer exchange or diafiltration
Housing MaterialRegenerated Cellulose (RC)
IncludesPolypropylene filtrate collection cup; sample filter cup; concentrate/retentate cup and cap
TypeCentrifugal Filters
Volume (Metric)15 to 70 mL
Unit SizePK