Search Thermo Fisher Scientific
Catalog Number | Length (Metric) | Diameter (Metric) Outer |
---|---|---|
FKS10653004 | 33.02 cm | 0.65 cm |
FIS13-678-4 | 14.6 cm | 0.65 cm |
FKS10528603 | 22.86 cm | 0.55 cm |
Corning disposable glass Pasteur pipets are available in convenient Packages. One-time use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents. These pipets feature a constriction one inch below the top to facilitate plugging. They are ideal for rapid nonvolumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.