Check Order Status
PFA Class a Volumetric Flask with Screwcap
Actual product may vary
Fisherbrand
PFA Class a Volumetric Flask with Screwcap
Manufactured from PerFluoroAlkoxy (PFA) and calibrated to contain (TC, In). Autoclavable and easy to clean.
Have Questions?
Catalog Number
Capacity (Metric)
FKS10642683
100 mL
Catalog number
FKS10642683
Price (SGD)
830.00
EA
Add to cart
Capacity (Metric):
100 mL
Price (SGD)
830.00
EA
Add to cart
PFA (PerFluoroAlkoxy) construction
PFA screw cap with molded sealing ring
Calibrated to contain (TC, In)
Autoclavable and easy to clean
Error limits according to class A
Compliant with DIN EN ISO 104
Includes one batch certificate
Exposure to temperatures up to 121°C (autoclaving) will not cause permanently exceeded error limits
Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions
Specifications
Certifications/Compliance
DIN EN ISO 104
Closure Material
PFA
Capacity (Metric)
100 mL
Material
PFA
Limit of Error
0.10 mL
Product Type
Volumetric flask
Autoclavable
Autoclavable
Class
A