Cimarec™ Power Direct High Capacity Stirrer
Cimarec™ Power Direct High Capacity Stirrer
Actual product may vary
Cimarec™ Power Direct High Capacity Stirrer
Cimarec™ Power Direct High Capacity Stirrer
Cimarec™ Power Direct High Capacity Stirrer
Fisherbrand
Cimarec™ Power Direct High Capacity Stirrer
Stir up to 40L with the integrated controller of the Thermo Scientific™ Cimarec™ Power Direct, ideal for high-capacity stirring.
Have Questions?
Catalog NumberCapacity
FKS1064240340 L
Catalog number FKS10642403
Price (SGD)
5,693.00
EA
Add to cart
Capacity:
40 L
Price (SGD)
5,693.00
EA
Add to cart
Stir up to 40L with the integrated controller of the Thermo Scientific™ Cimarec™ Power Direct, ideal for high-capacity stirring.
Features:
  • Quick Stop function stops the drive within two seconds, minimizing after runs and reducing the risk of breakage
  • Stir speeds from 100 to 2000rpm
  • Integrated controller has an adjustable power setting that decreases power consumption and lowers heat output
  • Sealed stainless-steel housing is easy to clean and chemical resistant
  • Operating conditions are -10° to +40°C
  • Protection Class IP22; easily cleaned with running water
Includes: All models are 100-240V and contain a cord set with various plugs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for 5 years thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Speed100 to 2000 rpm
Top Plate MaterialStainless Steel
Capacity40 L
Voltage100/240 V
Wattage5/10/15/20 w
Dimensions (L x W x H)11.6 x 9.4 x 1.4 in. (295 x 240 x 35 mm)
Height (English)1.4 in.
Height (Metric)35 mm
Length (English)11.6 in.
Length (Metric)295 mm
Width (English)9.4 in.
Width (Metric)240 mm
Certifications/ComplianceCE marked
Warranty and Service Offering90 Days (Limited)
ModelPower Direct
Protection ClassIP22
Temperature Range (Metric)-10°C to +40°C (Operating)
TypeStirrer
Weight (English)5.5 lb.
Weight (Metric)2.5 kg
Unit SizeEA