Plastipak™ Plastic Concentric Luer-Lock Syringe
Plastipak™ Plastic Concentric Luer-Lock Syringe
Actual product may vary
Fisherbrand
Plastipak™ Plastic Concentric Luer-Lock Syringe
Enables accurate dosing and visualisation of the syringe contents. BD Medical™ Plastipak™ Plastic Concentric Luer-Lock Syringe are each tested for sterility, pyrogenicity and toxicity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
FKS1063653150 mL
FKS1056921520 mL
Catalog number FKS10636531
Price (SGD)
235.00
EA
Add to cart
Volume (Metric):
50 mL
Price (SGD)
235.00
EA
Add to cart
  • Concentric Luer-Lok
  • Latex free stopper
  • CE marked
  • Polypropylene
  • Sterile
  • Disposable
  • Without needleFor needles, please see our Microlance™ disposable needles range. All products are for research use only, unless otherwise indicated.
Specifications
Certifications/ComplianceCE Mark, European Directive 93/42 / EEC (compliance procedure Annex V and VII) Certificates: No. 95 Q 252156 and No. 252231 Q (references 309661 & 309620 ) ; certified by NSAI (No. 0050)
MaterialPlastic
ColorTransparent
Diameter (Metric) Outer29.4 mm
Latex FreeYes
Length (Metric)133 mm
Product TypeConcentric Luer-lock Syringe
DisposableSingle-use
Graduations1 mL
QuantityPk. of 60
Volume (Metric)50 mL
Connection TypeConcentric Luer Lock