Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Size |
---|---|---|
FKS10633151 | 500 mL | Stopper No. 7 |
FKS10444881 | 250 mL | Stopper No. 6 |
4102-2000 also known as NLG4102-2000 | 2000 mL | Stopper No. 10 |
FIS10-041-10F | 2000 mL | Stopper No. 10 |
FKS10065200 | 2000 mL | Stopper No. 10 |
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.