1x1 image pixel for data collection
96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate
96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate
Actual product may vary
Fisherbrand
96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate
Color:Clear; Sterility:Non-sterile; Lid:Without Lid
Have Questions?
Catalog NumberColor
FKS10629151Clear
Catalog number FKS10629151
Price (SGD)
2,292.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Color:
Clear
Price (SGD)
2,292.00
PK
Add to cart
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
BarcodeAlphanumeric
FormatHalf-Area
LidWithout Lid
Array8 x 12
Binding TypeNot Treated
ColorClear
For Use With (Equipment)Plate readers
MaterialPolystyrene
No. of Wells96
Volume (Metric) Working25 to 125 μL
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
For Use With (Application)Suitable for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
No. per Case100/Cs.
No. per Pack25/Pk.
Quantity100/Cs.
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link