Nalgene™ PMP Griffin Low-Form Plastic Beakers
Nalgene™ PMP Griffin Low-Form Plastic Beakers
Actual product may vary
Nalgene™ PMP Griffin Low-Form Plastic Beakers
Nalgene™ PMP Griffin Low-Form Plastic Beakers
Nalgene™ PMP Griffin Low-Form Plastic Beakers
Fisherbrand
Nalgene™ PMP Griffin Low-Form Plastic Beakers
Continuously use these transparent, PMP beakers with easy-to-read, silk-screened graduation at temperatures of 150°C with intermittent service to 175°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS106227414000 mL
1203-0100100 mL
1203-0250250 mL
1203-0600600 mL
FIS02-591-15G600 mL
FIS02-591-15H1000 mL
FIS02-591-15AA4000 mL
Catalog number FKS10622741
Price (SGD)
185.00
EA
Add to cart
Capacity (Metric):
4000 mL
Price (SGD)
185.00
EA
Add to cart
Continuously use Thermo Scientific™ Nalgene™ PMP Griffin Low-Form Beakers at temperatures of 150°C with intermittent service to 175°C. These polymethylpentene beakers have the transparency of glass, excellent chemical resistance and heat tolerance. All sizes of beakers have easy-to-read, silk-screened graduations.
  • Transparent PMP material allows for a clear view of beaker contents
  • No-drip, single-spout design provides precision pouring control
  • With ribs for easy stacking
  • Silk screened with graduations, size code, resin code, catalog number, max. temperature and “No Flame” symbol for easy identification and safe use
  • Autoclavable/Graduated/Transparent
  • Meets ISO/DIN 7056 International Standards for Laboratory Plasticware Accuracy
.
Specifications
ColorClear
DescriptionBeaker Griffin Low-Form PMP 4000 mL
Capacity (Metric)4000 mL
MaterialPMP
Graduation Range500 to 4000 mL
Increments250 mL
Quantity1/Pk.; 4/Cs.
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Certifications/ComplianceISO/DIN 7056
Scale500mL to 4000ml in 250mL increments
StyleGriffin, Low Form
Unit SizeEA