DURAN™ GLS 80™ Pouring Ring, Blue PP
DURAN™ GLS 80™ Pouring Ring, Blue PP
Actual product may vary
DURAN™ GLS 80™ Pouring Ring, Blue PP
DURAN™ GLS 80™ Pouring Ring, Blue PP
Fisherbrand
DURAN™ GLS 80™ Pouring Ring, Blue PP
Blue PP GLS 80™ wide-mouth screw cap (plug seal) and drip free, PP pouring ring for GLS 80™ bottles
Have Questions?
Catalog NumberThread Size (Metric)
FKS10611724GLS 80
Catalog number FKS10611724
Price (SGD)
89.00
PK
Add to cart
Thread Size (Metric):
GLS 80
Price (SGD)
89.00
PK
Add to cart
Permits opening and closing of the DURAN™ GLS 80™ bottle with only a three-quarter turn. A matching PP pouring ring is also available, permitting clean, drip-free use.
  • Triple thread permits user friendly opening and closing with only a three-quarter turn
  • Cap offer a linerless, integral lip seal
  • Pouring ring for drip-free pouring and clean, safe working
Specifications
DescriptionPouring ring
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Sample collection; Culturing vessel; Storage of media and reagents; Transport and shipping
MaterialPolypropylene
Quantity10/Pk.
Thread Size (Metric)GLS 80
Neck Finish16 mm
AutoclavableAutoclavable
Diameter (Metric)87 mm
Unit SizePK