250 mL Vacuum Filter/Storage Bottle System
250 mL Vacuum Filter/Storage Bottle System
Actual product may vary
250 mL Vacuum Filter/Storage Bottle System
Vacuum filtration systems use a square filter membrane support system that increases filtration rates with printed funnels to identify membrane type and product number. The system consists of a polystyrene filter funnel joined by an adapter ring to a removable polystyrene storage bottle with a separate sterile polyethylene cap.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterial (Membrane)Pore Size
FKS10609141PES0.22 μm
FIS09-761-2Nylon0.2 μm
CRG431096PES0.22 μm
Catalog number FKS10609141
Price (SGD)
580.00
PK
Add to cart
Material (Membrane):
PES
Pore Size:
0.22 μm
Price (SGD)
580.00
PK
Add to cart
  • Sterile vacuum filtration system size 250 mL with 45 mm neck size
  • Funnel contains 49.5 x 49.5 mm square membrate filter
  • Receiver easily unscrews from unit and can be sealed with a plug cap for filtrate storag
  • Low liquid retention funnel design
  • Filters feature printing on the funnel for easy product identification
  • Angled hose connector simplifies vacuum line attachment
  • Receiver bottles feature easy grip sides for improved handling
  • Extra plastic storage bottles are available
  • Prefilters must be ordered separately
  • Individually packaged
  • Radiation sterilized and certified nonpyrogenic
  • Caps for receiver bottles are sterile and individually packaged
  • Units are color coded by membrane type and labeled with both membrane material and pore size
  • All membrane types are tissue culture compatible, and heat-sealed to the support grid to maximize flowrate, and reduce foaming and protein denaturization
  • Cellulose Acetate (CA) membrane for low protein binding
  • Nylon membrane for low extractables
  • Cellulose nitrate (CN) for general-purpose filtration
  • Polyethersulfone (PES) for fast flowrates and low protein binding
  • Nylon, Cellulose Nitrate and Polyetersulfone (PES) membranes are detergent-free
  • Not for parenteral use
Specifications
ConnectorsAngled Hose
DescriptionIncludes square membrane filter, graduated funnel and receiver of clear polystyrene, polyethylene neck adapter, orange HDPE plug seal
DisposableSingle-use
For Use With (Equipment)Bottle
MaterialPolystyrene
Material (Membrane)PES
Pore Size0.22 μm
SterilitySterile
Filtration Area19.6 cm2
For Use With (Application)Filtering and storing cell culture media, biological fluids and other aqueous solutions
GraduationsYes
Quantity12/Cs.
TypeVacuum Filter System
Volume (Metric)250 mL
Volume (Metric) Receiver250 mL
Unit SizePK