Check Order Status
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
Element Search
Structure Search
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis, Fisher BioReagents™
Have Questions?
Catalog Number
Quantity
Packaging
FKS10607633
200 mL
Poly Bottle
Catalog number
FKS10607633
Price (SGD)
147.00
EA
Add to cart
Quantity:
200 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
147.00
EA
Add to cart
Specifications
Absorbance
0.4 max. (0.1% solution) at 230nm, 0.1 max. (0.1% solution) at 280nm
Chemical Name or Material
Sodium Dodecyl Sulfate
Color
Colorless
Name Note
20% Solution
CAS
151-21-3
View more
SDS Solution, 20% Sodium Dodecyl Sulfate Solution, Molecular Biology/Electrophoresis
DNase, RNase, and protease-free premixed solution containing 20% SDS for protein extraction and DNA extraction procedures
Reduce preparation time by using this premade SDS solution
SDS is a commonly used detergent in protein purification, polyacrylamide gel electrophoresis (PAGE), and other molecular biology applications
SDS is also used to disrupt cell membranes and dissociate nucleic acid-protein complexes in DNA extraction procedures
Intended for research use only; not for human or animal diagnostic purposes