Percoll™ Centrifugation Media
Percoll™ Centrifugation Media
Actual product may vary
Fisherbrand
Percoll™ Centrifugation Media
For separation of cells, subcellular particles, and larger viruses (down to approx. 70S) under gentle conditions that preserve viability and morphological integrity. Cytiva™ Percoll™ Centrifugation Media Percoll is composed of colloidal silica coated with polyvinylpyrrolidone (PVP).
Have Questions?
Catalog NumberQuantity
FKS106070951 L
Catalog number FKS10607095
Price (SGD)
-
Quantity:
1 L
  • Gradients can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor
  • Low viscosity of media allows cell isolations on preformed gradients to be completed in only a few minutes using low centrifugal forces (200 to 1000xG)
  • Iso-osmotic gradients throughout cover a range of densities up to 1.3g/mL
  • Nontoxic, almost chemically inert and does not adhere to membranes
  • Adjustable to physiological ionic strength and pH; can be buffered within pH range 5.5 to 10.0 without change in properties
  • Percoll is a silica sol with nondialyzable PVP coating; 15-30nm dia. bead size
For in vitro research use only.
Specifications
Quantity1 L
For Use With (Application)Cell Isolation
Viscosity15cP
Osmolality< 25 mOsm/kg
ColorColorless to Slightly Yellow
pH8.5 to 9.5
Product TypeCentrifugation Media
Shelf Life5 years
Density1.135 g/mL
FormLiquid
Unit SizeEA
Contents & Storage
4–30°C