Stripette™ Paper/Plastic-Wrapped Disposable Polystyrene Serological Pipettes, Sterile
Stripette™ Paper/Plastic-Wrapped Disposable Polystyrene Serological Pipettes, Sterile
Actual product may vary
Fisherbrand
Stripette™ Paper/Plastic-Wrapped Disposable Polystyrene Serological Pipettes, Sterile
Manufactured with optically clear polystyrene for accurate measurement. Corning™ Stripette™ Individually Wrapped Plastic or Paper/Plastic Disposable Polystyrene Serological Pipets are ideal for sterile tissue culture applications, especially while wearing latex gloves, as the wrap reduces static cling.
Have Questions?
Catalog NumberVolume (Metric)Color
FKS1060615125 mLRed
Catalog number FKS10606151
Price (SGD)
547.00
PK
Add to cart
Volume (Metric):
25 mL
Color:
Red
Price (SGD)
547.00
PK
Add to cart
  • Sterile
  • RNase-/DNase-free
  • Nonpyrogenic
  • Noncytotoxic
  • BSE/TSE-free
  • Exclusive anti-drip tip assures accurate delivery
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Bi-directional graduations provide choice of ascending and descending scales
  • Negative graduations allow additional working volume
  • Three packaging options: Individually wrapped, paper/plastic; Individually wrapped, clear plastic; Bulk packed
  • Volumetric accuracy of ±2% at full capacity
Specifications
Accuracy±2%
Certifications/ComplianceNon-pyrogenic, DNase, RNase-free
MaterialPolystyrene
SterilitySterile
GraduatedYes
Graduations0.2 mL
Length (English)13.54 in.
Negative Graduations10 mL
Quantity200/Cs.
Tip StyleRound
Tolerance±2%
Packaging TypeIndividually Wrapped, Paper/Plastic Packaging
ColorRed
Volume (Metric)25 mL
Length (Metric)34.4 cm
TypeSerological Pipet