DURAN™ Original Laboratory GL Bottle Pouring Ring
DURAN™ Original Laboratory GL Bottle Pouring Ring
Actual product may vary
Fisherbrand
DURAN™ Original Laboratory GL Bottle Pouring Ring
Screw cap with reliable plug seal.
Have Questions?
Catalog NumberThread Size (Metric)Color
FKS10602981GL 45Blue
Catalog number FKS10602981
Price (SGD)
28.00
PK
Add to cart
Thread Size (Metric):
GL 45
Color:
Blue
Price (SGD)
28.00
PK
Add to cart
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
  • Cap with integral lip seal provides effective sealing for short term storage
  • Pouring ring for drip-free pouring
  • The colour helps prevent interchanging of cap mix-ups and reduces the risk of cross contamination
  • Fully compatible with all the GL 45 threads and therefore highly versatile
Specifications
ColorBlue
DescriptionPouring ring
For Use With (Application)Autoclaving media and reagents; Short term sample and media storage; color coding and identification
MaterialPolypropylene
Quantity10/Pk.
Thread Size (Metric)GL 45