Shaker Vibramax 100
Shaker Vibramax 100
Actual product may vary
Fisherbrand
Shaker Vibramax 100
Compact, space-saving shaker/mixer. The Shaker Vibramax 100 has a load capacity of 2 kg.
Have Questions?
Catalog number FKS10588751
Price (SGD)
5,812.00
EA
Add to cart
Price (SGD)
5,812.00
EA
Add to cart
  • The 3 mm vibration orbit gives excellent results especially in large vessels – allows for smooth to ridged mixing of viscous media and solids
  • Set and continuously adjust the variable speed on the analog control knob from 150 to 1,350 rpm
  • Optional attachment with tension rollers accepts any glassware size
  • A large selection of attachments is provided for the use of various vessels and sizes
  • An analog process timer allows for unattended operation and can be set from 1 to 120 minutes. When the set time has elapsed an acoustic alarm will sound and the operation stops
Specifications
Orbit3 mm
Load Bearing Capacity (Metric)2 kg
Speed Range150 to 1350 rpm
Electrical Requirements230 V, 31 W
Product TypeShaker
Load Bearing Capacity (English)4.4 lb.
Certifications/ComplianceDIN EN 60529, IP 30
Dimensions (LxW) Platform220 x 220 mm
ModelVibramax 100
IncludesTimer, overheat protection
Timer Range1 to 120 min.
Unit SizeEA