Flameboy Gas Cartridge Adapter
Flameboy Gas Cartridge Adapter
Actual product may vary
Fisherbrand
Flameboy Gas Cartridge Adapter
Use this adaptor with Flameboy™ Sterilizing Pistols. Integra Biosciences™ Gas Cartridge Adapters are easily mounted via an intelligent click and go mechanism.
Have Questions?
Catalog number FKS10586132
Price (SGD)
439.00
EA
Add to cart
Price (SGD)
439.00
EA
Add to cart
  • Adapter with quick coupling for Campingaz CV360 gas cartridge
  • For use with Flameboy Sterilizing Pistols
Specifications
Product TypeAdapter
For Use With (Equipment)Campingaz CV 360
Unit SizeEA