1700 Series Gastight™ Syringes: N Termination
1700 Series Gastight™ Syringes: N Termination
Actual product may vary
Fisherbrand
1700 Series Gastight™ Syringes: N Termination
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Needle GaugeNeedle Point Style
FKS1057774150 μL22s G2
FKS10615061100 μL22s G2
FIS14-815-82100 μL22s G5
Catalog number FKS10577741
Price (SGD)
295.00
EA
Add to cart
Volume (Metric):
50 μL
Needle Gauge:
22s G
Needle Point Style:
2
Price (SGD)
295.00
EA
Add to cart
Gastight version of the original 700 series syringes, ideal for dispensing volumes from 1 up to 500μL
Syringe series offers a variety of different terminations and needle options.
1700 Series Syringe Terminations
  • Cemented Needle (N)- For low volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
  • Needle Point Styles (Point styles 4 and AS are available on request)
    • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34-10
    • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauges: 34-10
    • Point Style 4 - Standard 12° beveled needle point is recommended for life science applications. Available gauges: 34-10. Special needle angles such as 0° to 60° available upon request.
    • Point Style 5 - Conical needle with side port for penetration of septa, thin-gauged vinyls and plastics without coring. Available gauges: 26-10
    • Point Style AS - Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration. Available gauges: 26-10
Specifications
AutoclavableNot Autoclavable
MaterialBorosilicate Glass (Barrel), PTFE (Tip), Stainless Steel (Plunger)
Needle Gauge22s G
Diameter (English) Inner0.04 in.
Diameter (English) Inner Needle0.0059 in.
Diameter (Metric) Inner Needle0.15 mm
Diameter (English) Outer Needle0.0283 in.
Diameter (English) Outer0.3 in.
Diameter (Metric) Inner1.03 mm
Diameter (Metric) Outer7.75 mm
Diameter (Metric) Outer Needle0.72 mm
For Use With (Application)Ideal for dispensing both liquids and gases, useful for samples with dissolved or suspended molecules
Length (English)2 in.
Length (English) Needle2 in.
Length (Metric)51 mm
Product TypeGastight Syringe
Max. Pressure1000 psi
Max. Temperature (English)122°F
Max. Temperature (Metric)50°C
Min. Temperature (English)50°F
Model1705
Needle Point Number2
Min. Temperature (Metric)10°C
QuantityEa.
Volume (Metric)50 μL
Needle Point Style2
Connection TypeCemented
Unit SizeEA