PTFE Tall-form Evaporating Dish with Spout
PTFE Tall-form Evaporating Dish with Spout
Actual product may vary
Fisher Catalogue
PTFE Tall-form Evaporating Dish with Spout
Reduce spills when pouring excess liquid with integrated molded spout. Cowie Technology™ PTFE Tall-form Evaporating Dish with Spout includes a super-smooth surface for \maximum water and solvent evaporation.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FKS10577282100 mL64 mm50 mm
Catalog number FKS10577282
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
100 mL
Diameter (Metric):
64 mm
Height (Metric):
50 mm
  • Isostatically molded in pure PTFE
  • Chemically resistant and inert
  • Super smooth internal finish
  • Usable to 280°C
  • Molded spout for easy pouring
Specifications
Capacity (English)3.33 fl oz.
Bottom ShapeFlat
Capacity (Metric)100 mL
MaterialPTFE
ShapeRound
TypeEvaporating Dish
IncludesSpout
Interior FinishSmooth
Temperature Range (Metric)-200°C to +280°C
Height (Metric)50 mm
Diameter (Metric)64 mm
Unit SizeEA