Sterileware™ Sterile Styrene Sampler Spoons
Sterileware™ Sterile Styrene Sampler Spoons
Actual product may vary
Fisher Catalogue
Sterileware™ Sterile Styrene Sampler Spoons
Polystyrene plastic sterile sampler spoons are designed to gather small amounts of sample material.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FKS105739241.23 mL200/Cs.
FIS14-429D14.79 mL200/Cs.
FIS14-429B2.46 mL200/Cs.
FIS14-429A1.23 mL200/Cs.
FIS14-429C4.93 mL200/Cs.
Catalog number FKS10573924
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1.23 mL
Quantity:
200/Cs.
Manufactured from polystyrene plastic. SP Scienceware™ Sterileware™ Sterile Styrene Sampler Spoons are designed to gather small amounts of sample material.
  • Sterile, individually packaged, sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Processing for Sterilization
  • FDA grade materials; safe for food, drug and cosmetic contact
  • Long handle reaches easily into jars, bottles and other confined areas, and minimizes the potential for personal contact with sampled material
  • Overall length 17.8 cm (7 in.)
Specifications
Product TypeSpoon
MaterialPolystyrene
AutoclavableNot Autoclavable
IncludesCertificate of sterilization
Handle TypeLong
Capacity (English)0.25 oz.
Capacity (Metric)1.23 mL
DisposableSingle-use
SterilitySterile
ColorWhite
Certifications/ComplianceFDA
For Use With (Application)Designed to gather small amounts of sample material
Quantity200/Cs.
Dimensions (L x W)17.8 x 2 cm
Length (English)7 in.
Length (Metric)17.8 cm
Width (English)0.78 in.
Width (Metric)2 cm
Unit SizePK