Borosilicate Glass Class B Burette with Stopcock
Borosilicate Glass Class B Burette with Stopcock
Actual product may vary
Fisherbrand
Borosilicate Glass Class B Burette with Stopcock
Manufactured from borosilicate glass and omplies with BS-ISO 385 standards. Fisherbrand™ Borosilicate Glass Class B Burette with Stopcock includes single bore, interchangeable glass or PTFE stopcock with 0.10mL blue ceramic graduations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberStopcock Material
FKS10569771PTFE
FKS10448972Glass
Catalog number FKS10569771
Price (SGD)
106.00
EA
Add to cart
Stopcock Material:
PTFE
Price (SGD)
106.00
EA
Add to cart
Manufactured from borosilicate glass and omplies with BS-ISO 385 standards. Fisherbrand™ Borosilicate Glass Class B Burette with Stopcock includes single bore, interchangeable glass or PTFE stopcock with 0.10mL blue ceramic graduations.
  • Manufactured from borosilicate glass
  • Complies with BS-ISO 385
  • Single bore
  • Interchangeable glass or PTFE stopcock
  • 0.10mL blue ceramic graduations
Specifications
Capacity (Metric)50 mL
MaterialBorosilicate Glass
Graduations0.1 mL
IncludesSingle Bore, Interchangeable Stopcock
ScaleBlue
Stopcock MaterialPTFE
Tolerance±0.1 mL
TypeBuret
ClassClass B
Unit SizeEA