UV-Cuvettes - CERTIFIED LIFE SCIENCE QUALITY
Actual product may vary
Fisherbrand
UV-Cuvettes - CERTIFIED LIFE SCIENCE QUALITY
BRAND™ disposable UV-cuvettes are a cost-effective, durable alternative to fragile quartz cuvettes in numerous applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
FKS105665811.5 to 3 mL100 Pack
FIS03-840-30070 to 850 μL100 Pack, Individually Wrapped
FIS13-688-731.5 to 3 mL500 Pack
FIS13-688-741.5 to 3 mL100 Pack
FIS13-878-12170 to 850 μL100 Pack
FKS1029027270 to 850 μL100 Pack
FIS13-878-12370 to 550 μL100 Pack
FIS13-878-12470 to 550 μL500 Pack
FIS14-377-0092.5 to 4.5 mL100 Pack
FKS100467312.5 to 4.5 mL100 Pack
Catalog number FKS10566581
Price (SGD)
227.00
Pack
Capacity (Metric):
1.5 to 3 mL
Quantity:
100 Pack
Price (SGD)
227.00
Pack
BrandTech™ BRAND™ UV-Cuvette Disposable Cuvets provide transparency from 230 to 900nm and eliminate cleaning and risk of cross-contamination. Ideal for DNA, RNA and protein analysis. Compatible with with most polar organic solvents, acids, and bases.
CERTIFIED LIFE SCIENCE QUALITY cuvettes from BRAND™ are manufactured from virgin raw materials under controlled room conditions, with intensive in-process controls, visual inspections, batch management, and final product inspection to ensure proven functionality.
  • Advanced cuvette design includes recessed windows to protect against scratching in the optical pathway
  • Disposable - cleaning and cross-contamination risks are eliminated
  • Ideal for DNA, RNA, and protein analysis
  • Chemical resistant - can be used with most polar organic solvents, acids, and bases
  • Also compatible with acetone, butanone, DMF, and concentrated hydrochloric acid
  • Grouped by manufacturing mold cavity to ensure lowest variation in extinction coefficient
  • Fits most spectrophotometers without adapters
Ultramicro
  • Available with 8.5 and 15 mm window heights (Z-dimension)
  • May be used with sample volumes as small as 70 μL
  • Can be fitted with plug seal caps (sold separately) for sample storage
  • Individually wrapped UV-Cuvettes are DNA-, DNase-, and RNase-free
Semimicro
  • For sample volumes between 1.5 and 3.0 mL
Macro
  • For sample volumes between 2.5 and 4.5 mL
Specifications
SterilityNon-sterile
TypeSemi-Micro Cuvette
ColorClear
DisposableSingle-use
Capacity (Metric)1.5 to 3 mL
MaterialPlastic
IncludesCuvette Only
Quantity100 Pack
Path Length10 mm
Wavelength Range230 to 900 nm
Height (Metric)45 mm
For Use With (Equipment)Spectrophotometers, Photometers
Width (Metric)12.5 mm
For Use With (Application)DNA, RNA, Protein Analysis, Molecular Biology
Lightpath10 mm
ShapeRectangle
Manufacturing QualityCERTIFIED LIFE SCIENCE QUALITY
Unit SizePack