Screw Vial Convenience Kit, solid-top cap, 4ml clear glass vial
Screw Vial Convenience Kit, solid-top cap, 4ml clear glass vial
Actual product may vary
Fisherbrand
Screw Vial Convenience Kit, solid-top cap, 4ml clear glass vial
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Storage vials range from 2mL to 40mL.
Have Questions?
Catalog NumberColor
FKS10560116Clear
Catalog number FKS10560116
Price (SGD)
202.00
PK
Add to cart
Color:
Clear
Price (SGD)
202.00
PK
Add to cart
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
Specifications
DescriptionScrew Vial Convenience Kit, Solid-top Cap
ColorClear
MaterialGlass
Volume (Metric)4 mL
No. per Pack100
Closure TypeScrew Cap, Solid Top
Unit SizePK