700/1700 Series Microliter/Gastight™ Syringes
700/1700 Series Microliter/Gastight™ Syringes
Actual product may vary
Fisherbrand
700/1700 Series Microliter/Gastight™ Syringes
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry. Each is manufactured to achieve the highest level of accuracy and precision possible.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Model
FKS1055020310 μL1701
FKS10485952100 μL1710
Catalog number FKS10550203
Price (SGD)
306.00
PK
Add to cart
Volume (Metric):
10 μL
Model:
1701
Price (SGD)
306.00
PK
Add to cart
Specifications
AutoclavableAutoclavable
MaterialBorosilicate Glass (Barrel), PTFE (Tip), Stainless Steel (Plunger)
Diameter (English) Inner0.018 in.
Diameter (English) Outer0.26 in.
Diameter (Metric) Inner0.46 mm
Diameter (Metric) Outer6.6 mm
For Use With (Application)Provides unprecedented functionality for controlled animal injections
Product TypeGastight Syringe
Max. Pressure1000 psi
Max. Temperature (English)239°F
Max. Temperature (Metric)115°C
Min. Temperature (English)50°F
Model1701
Needle Point NumberNo Needle Included
Min. Temperature (Metric)10°C
QuantityEa.
Volume (Metric)10 μL
Connection TypeRemovable
Unit SizePK