Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Actual product may vary
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
Fisherbrand
Nalgene™ Autoclavable Polypropylene Desiccators: Blue body with clear polycarbonate cover
View contents easily with these autoclavable polypropylene desiccators, featuring a transparent PC cover and robust body resistant to all solid desiccants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Diameter (Metric) Inner
FKS10548842149 mm149 mm
FIS08-642262 mm246 mm
FIS08-642-15149 mm149 mm
Catalog number FKS10548842
Price (SGD)
225.00
EA
Add to cart
Height (Metric):
149 mm
Diameter (Metric) Inner:
149 mm
Price (SGD)
225.00
EA
Add to cart
View contents easily with Thermo Scientific™ Autoclavable Polypropylene Desiccators, featuring a robust body and high resistance to all solid desiccants. These desiccators have a transparent polycarbonate cover. The small size (5315-0150) comes with a 140 mm aluminum plate. The plate for the large size (5309-0250) is sold separately.
  • Clear polycarbonate (PC) cover has recessed rim to keep grease off benchtop when cover is set down
  • No gasket required, only grease is needed
  • Large desiccator (Cat. No. 5310-0250) takes any standard 230mm desiccator plate such as Cat. No. 5312-0230 (sold separately)
  • Autoclavable; repeated autoclaving will reduce the life of the product
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
AutoclavableAutoclavable
DescriptionDesiccator PP/PC 150mm
Cabinet StyleTop Loading
MaterialPolypropylene, Polycarbonate
Max. Clearance Above Plate3.4 in.
No. of Shelves1
No. per Pack1
Height (English)5.88 in.
Height (Metric)149 mm
Diameter (English) Inner5.87 in.
Diameter (English) Outer7.125 in.
Diameter (Metric) Inner149 mm
Diameter (Metric) Outer181 mm
Unit SizeEA