1x1 image pixel for data collection
Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Actual product may vary
Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Fisherbrand
Nalgene™ Wash Bottles made with Teflon™ fluoropolymer
Ensure contamination-free dispensing with these chemical-resistant wash bottles made with Teflon™ fluoropolymer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS10548701250 mL24 mm
FIS03-409-12A125 mL24 mm
FIS03-409-12C250 mL24 mm
2403-0250250 mL24 mm
2403-0500500 mL38-430 mm
FIS03-409-12E500 mL38-430 mm
FKS10170551500 mL38-430 mm
Catalog number FKS10548701
Price (SGD)
591.00
EA
Add to cart
Capacity (Metric):
250 mL
Closure Size:
24 mm
Price (SGD)
591.00
EA
Add to cart
Ensure contamination-free dispensing with Thermo Scientific™ Nalgene™ Wash Bottles made with Teflon™ fluoropolymer, useful for trace element work and other high-purity applications. These phthalate-free bottles offer excellent chemical resistance and are molded entirely of fluoropolymer. Designed to dispense through the top with a closure and stem molded in one piece for reliable service. The precision-molded tip provides a straight, steady stream and ultimate control without sputtering.
  • With ETFE screw closure/stem and draw tube
  • Demonstrates excellent chemical resistance to nearly any chemical in your lab including solvents—won't become brittle or crack
  • Inert fluoropolymer construction is low in extractables and low binding—useful for trace element work
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Autoclavable/Transparent
  • Sputter-free, precision stream
.
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.
Specifications
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
ColorTranslucent
MaterialFEP
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
MouthNarrow
ShapeCylindrical
Height (Metric)178 mm
Product TypeWash Bottle
Height (English)7 in.
Closure MaterialETFE
Closure Size24 mm
Diameter (Metric)60.2 mm
Closure TypeScrew Cap
Through the Closure DispensingYes
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link