Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Socket Joint |
|---|---|---|
| FKS10538190 | 100 mL | 24/29 |
| FKS10488560 | 1000 mL | 29/32 |
| FKS10829580 | 100 mL | 14/23 |
| FKS10671071 | 150 mL | 24/29 |
| FKS10765111 | 25 mL | 14/23 |
| FKS10558590 | 25 mL | 24/29 |
| FKS12682002 | 250 mL | 24/29 |
| FKS10488570 | 500 mL | 24/29 |
| FKS10809900 | 50 mL | 14/23 |
| FKS10181780 | 50 mL | 24/29 |