96-Well, Cell Culture-Treated, Flat-Bottom Microplate
96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Actual product may vary
Fisher Catalogue
96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Black, Clear Bottom; Sterility: Sterile; Lid: With Lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging
FKS10530753Individually wrapped, 48/Cs.
FIS07-200-565Individually wrapped, 48/Cs.
Catalog number FKS10530753
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Packaging:
Individually wrapped, 48/Cs.
  • Bottoms are 60% thinner than conventional polystyrene microplates, resulting in lower background fluorescence and enabling readings down to 340 nm.
  • Opaque walls prevent well-to-well cross-talk.
  • Optically clear flat bottom permits direct microscopic viewing.
  • Specifications
    FormatStandard
    LidWith Lid
    Culture Area0.32 cm2
    Array8 x 12
    PackagingIndividually wrapped, 48/Cs.
    ColorBlack, Clear Bottom
    MaterialPolystyrene
    No. of Wells96
    Volume (Metric) Working75 to 200 μL
    SterilitySterile
    Surface TreatmentCell Culture
    Well ShapeFlat Bottom (F-well)
    For Use With (Application)Attachment and growth of anchorage-dependent cells
    No. per Case48/Cs.
    No. per Pack1/Pk.
    Quantity48/Cs.
    Unit SizePK