Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A
Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A
Actual product may vary
Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A
Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A
Fisherbrand
Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A
Combines high loading capacity with fine particle retention. Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A are ideal for gravimetric determination of airborne particulates, stack sampling and absorption methods of air pollution monitoring.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS10529441125 mm
FKS1031787160 mm
Catalog number FKS10529441
Price (SGD)
734.00
PK
Add to cart
Diameter (Metric):
125 mm
Price (SGD)
734.00
PK
Add to cart
Combines high loading capacity with fine particle retention. Whatman™ Binder-Free Glass Microfiber Filters, Grade GF/A are ideal for gravimetric determination of airborne particulates, stack sampling and absorption methods of air pollution monitoring.
Whatman glass microfiber filters without binder feature low resistance to fluid flow and high particle-loading capacity. These filters can be used at temperatures up to 500°C. The fine capillary structure of glass fiber filters can absorb large quantities of water, enabling spot tests and liquid scintillation counting methods.
Glass microfiber filters are mainly used in applications such as air or water monitoring and can be used as pre-filters for membranes.
  • High flow rate
  • Good loading capacity
  • Water pollution effluent monitoring
  • Filtering water, algae, bacterial cultures
  • Foodstuff analyses
  • Protein filtration
  • Radioimmunoassay of weak β-emitters
Specifications
DescriptionGE’s Whatman Grade GF/A glass microfiber filters feature fine retention. GF/A filters are suitable for water quality parameter measurement and air filtration applications.
FormatDisc
MaterialBorosilicate Glass
Particle Retention1.6 μm
Flow Rate13 sec./100mL
For Use With (Application)Used for high-efficiency general purpose laboratory filtration, including water pollution monitoring of effluents, for filtration of water, foodstuff analyses, protein filtration, and radioimmunoassay of weak β emitters.
GradeGrade GF/A
Quantity100 Pieces
ShapeCircle
Temperature (English) Max.932°F
Temperature (Metric) Max500°C
Thickness (English)0.01 in.
Thickness (Metric)260 μm
TypeFine Retention Filter
Waterflow Rate143 mL/min
Diameter (English)4.92 in.
Diameter (Metric)125 mm
Unit SizePK